E-PAK™ Cartridges, SiliaMetS™ Thiol
E-PAK™ Cartridges, SiliaMetS™ Thiol
Actual product may vary
Thermo Scientific Chemicals
E-PAK™ Cartridges, SiliaMetS™ Thiol
E-PAK™ Cartridges, SiliaMetS™ Thiol (Si-Thiol) is our most versatile and robust metal scavenger for a variety of metals under a wide range of conditions conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Have Questions?
Catalog NumberDimensions (L x W)Quantity
ALF011821.0925 x 5 cm1 Pk.
Catalog number ALF011821.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Dimensions (L x W):
25 x 5 cm
Quantity:
1 Pk.
Metals removed:
  • Best scavenger: Ag, Hg, Os, Pd, and Ru
  • Good scavenger: Cu, Ir, Pb, Rh, Se, and Sn
Additional Information:
  • To facilitate the evaluations of small samples, we can provide loose media and lab scale cartridges on a custom basis.
  • Testing with loose media can be done with samples as small as a few milliliters and is normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
  • Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
  • Lab cartridges can be operated with a peristaltic pump or a small pressure vessel.
Specifications
TypeLab Scale E-PAK Cartridge
For Use With (Application)Lower residual metal concentration in Ag, Hg, Os, Pd, Ru
ColorWhite to yellow
Dimensions (L x W)25 x 5 cm
Quantity1 Pk.
pH2 to 9
Particle Size40 to 63 μm
SorbentThiol
Sorbent Weight200 g
Unit SizeEA