Ruthenium(III) chloride oxide, ammoniated
Ruthenium(III) chloride oxide, ammoniated
Ruthenium(III) chloride oxide, ammoniated
Ruthenium(III) chloride oxide, ammoniated
Thermo Scientific Chemicals
Ruthenium(III) chloride oxide, ammoniated
Microscopic stain | CAS: 11103-72-3 | Cl6H46N14O2Ru3 | 790.374 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALF011806.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialRuthenium(III) chloride oxide
Melting Point>500°C (decomposition)
Name NoteAmmoniated
CAS11103-72-3
ColorRed
View more
Microscopic stainRuthenium(III) chloride oxide, ammoniated is used as a reagent on plant material for staining pectin, gum, animal tissues and bacteria. It is also used in histology to stain aldehyde fixed mucopolysaccharides. Further, it serves as a pharmacological tool to study specific cellular mechanisms. It serves as an inhibitor of mitochondrial calcium uniporter. In addition, it is involved in reducing airway inflammation arises by pepper spray.