Ammonium iodide, ACS, 99.0% min
Ammonium iodide, ACS, 99.0% min
Ammonium iodide, ACS, 99.0% min
Ammonium iodide, ACS, 99.0% min
Thermo Scientific Chemicals
Ammonium iodide, ACS, 99.0% min
CAS: 12027-06-4 | H4IN | 144.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF011668.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium iodide
Melting Point551°C (sublimation)
CAS12027-06-4
ColorWhite to Yellow
Health Hazard 1H315-H319-H335
View more
Appropriately manufactured ammonium iodide is used in pharmaceutical industries. It is used in photographic chemicals, some medications and several chemical industries.