Copper(I) iodide, 98%
Copper(I) iodide, 98%
Copper(I) iodide, 98%
Copper(I) iodide, 98%
Thermo Scientific Chemicals
Copper(I) iodide, 98%
CAS: 7681-65-4
Have Questions?
Catalog NumberQuantity
ALF011606.30250 g
Catalog number ALF011606.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Boiling Point∼1290°C
Chemical Name or MaterialCopper(I) iodide
Melting Point605°C
CAS7681-65-4
ColorBrown
View more
It is used as a catalyst in organic synthesis, cathode ray tube coating, cloud seeding, and as temperature indicator with HgI2. It catalyzes the conversion of aryl, heteroaryl, and vinyl bromides into the corresponding iodides, which are employed in coupling reactions such as Suzuki, Stille, Sonogashira, Heck and Ullmann reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • As a catalyst in organic synthesis, cathode ray tube coating, temperature indicator with HgI2
RUO – Research Use Only