Ammonium dihydrogen phosphate, ACS, 98.0% min
Ammonium dihydrogen phosphate, ACS, 98.0% min
Ammonium dihydrogen phosphate, ACS, 98.0% min
Ammonium dihydrogen phosphate, ACS, 98.0% min
Thermo Scientific Chemicals
Ammonium dihydrogen phosphate, ACS, 98.0% min
CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Have Questions?
Catalog NumberQuantity
ALF011598.36500 g
Catalog number ALF011598.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium dihydrogen phosphate
Melting Point190°C
CAS7722-76-1
Health Hazard 1H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
View more
Preparation of baking powder with sodium bicarbonate, in fermentation.Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.