Potassium hydrogen phosphate, ACS, 98.0% min
Potassium hydrogen phosphate, ACS, 98.0% min
Potassium hydrogen phosphate, ACS, 98.0% min
Potassium hydrogen phosphate, ACS, 98.0% min
Thermo Scientific Chemicals
Potassium hydrogen phosphate, ACS, 98.0% min
CAS: 7758-11-4 | HK2O4P | 174.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number ALF011593.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium hydrogen phosphate
CAS7758-11-4
ColorWhite
Health Hazard 1H319: Causes serious eye irritation.
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. This material and/or its container must be disposed of as hazardous waste.
View more
It is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.