Niobium(V) chloride, 99.9% (metals basis)
Niobium(V) chloride, 99.9% (metals basis)
Niobium(V) chloride, 99.9% (metals basis)
Niobium(V) chloride, 99.9% (metals basis)
Thermo Scientific Chemicals
Niobium(V) chloride, 99.9% (metals basis)
CAS: 10026-12-7 | Cl5Nb | 270.156 g/mol
Have Questions?
Catalog NumberQuantity
ALF011548.22100 g
Catalog number ALF011548.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point248.2°C
Chemical Name or MaterialNiobium(V) chloride
Melting Point208.3°C
CAS10026-12-7
ColorWhite
View more
It is the precursor in the preparation of pure niobium, and ferroniobium. It is used as Lewis acid catalyst in the organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.