Silver foil, 0.025mm (0.001in) thick, hard, Premion™, 99.998% (metals basis)
Silver foil, 0.025mm (0.001in) thick, hard, Premion™, 99.998% (metals basis)
Silver foil, 0.025mm (0.001in) thick, hard, Premion™, 99.998% (metals basis)
Silver foil, 0.025mm (0.001in) thick, hard, Premion™, 99.998% (metals basis)
Thermo Scientific Chemicals
Silver foil, 0.025mm (0.001in) thick, hard, Premion™, 99.998% (metals basis)
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 250 mm
25 x 25 mm
50 x 50 mm
Catalog number ALF011498.GB
Price (MYR)
2,318.00
EA
Add to cart
Quantity:
50 x 250 mm
Request bulk or custom format
Price (MYR)
2,318.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilver foil
Name Note0.025mm (0.001 in.) thick, Hard, Temper: as rolled
CAS7440-22-4
Recommended StorageAmbient temperatures
Quantity50 x 250 mm