Ammonium hexafluorophosphate, 99.5%
Ammonium hexafluorophosphate, 99.5%
Ammonium hexafluorophosphate, 99.5%
Ammonium hexafluorophosphate, 99.5%
Thermo Scientific Chemicals
Ammonium hexafluorophosphate, 99.5%
Ammonium hexafluorophosphate, 99.5%, Alfa Aesar, Quantity: 100g, CAS: 16941-11-0, Melting Point: 198 deg.C, Molecular Formula: F6H4NP, Molecular Weight: 163.003 | CAS: 16941-11-0 | F6H4NP | 163.003 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF011495.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium hexafluorophosphate
Melting Point198°C
CAS16941-11-0
Health Hazard 1H314
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Ammonium hexafluorophosphate is used as a preservative. It is used as a starting material in the preparation of other hexafluorophosphates. Further, it is used in synthesis of organometalic compounds, especially rhodicinium hexafluorophosphate, which has inert and non-coordinating counter ion.