Tetrafluoroboric acid, 48% min w/w aq. soln.
Tetrafluoroboric acid, 48% min w/w aq. soln.
Tetrafluoroboric acid, 48% min w/w aq. soln.
Tetrafluoroboric acid, 48% min w/w aq. soln.
Thermo Scientific Chemicals
Tetrafluoroboric acid, 48% min w/w aq. soln.
CAS: 16872-11-0
Have Questions?
Catalog NumberQuantity
ALF011484.36500 g
Catalog number ALF011484.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrafluoroboric Acid
CAS16872-11-0
Boiling Point130°C (decomposition)
Health Hazard 1H314-H360FD
Density1.4
View more
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and treat the surface of substrates. It is used as radiotracers.