Silver wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis)
Silver wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis)
Silver wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis)
Silver wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis)
Thermo Scientific Chemicals
Silver wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis)
Form: Approx. 8.24g/m | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 cm
50 cm
Catalog number ALF011466.CD
Price (MYR)
4,237.00
EA
Add to cart
Quantity:
1000 cm
Request bulk or custom format
Price (MYR)
4,237.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilver
ColorSilver
CAS7440-22-4
AppearanceSilver-colored wire