Silver wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis)
Silver wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis)
Silver wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis)
Silver wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis)
Thermo Scientific Chemicals
Silver wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis)
Form: approx. 32.94g/m | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF011465.BU25 cm
Catalog number ALF011465.BU
Price (MYR)
755.00
EA
Add to cart
Quantity:
25 cm
Request bulk or custom format
Price (MYR)
755.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilver
ColorSilver
CAS7440-22-4
AppearanceSilver-colored wire