Silver wire, 0.5mm (0.02in) dia, 1/2 hard, 99.9% (metals basis)
Silver wire, 0.5mm (0.02in) dia, 1/2 hard, 99.9% (metals basis)
Silver wire, 0.5mm (0.02in) dia, 1/2 hard, 99.9% (metals basis)
Silver wire, 0.5mm (0.02in) dia, 1/2 hard, 99.9% (metals basis)
Thermo Scientific Chemicals
Silver wire, 0.5mm (0.02in) dia, 1/2 hard, 99.9% (metals basis)
Form: Approx. 2.06g/m, Temper: 1/2 hard | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF011433.G22 m
Catalog number ALF011433.G2
Price (MYR)
385.00
EA
Add to cart
Quantity:
2 m
Request bulk or custom format
Price (MYR)
385.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilver
ColorSilver
Name Note1/2 Hard
CAS7440-22-4
AppearanceSilver-colored wire