Form: approx. 3.5g/25 x 25mm | CAS: 7439-88-5 | Ir | 192.22 g/mol
Have Questions?
Catalog Number
Quantity
ALF011427.FF
25 x 25 mm
Catalog number ALF011427.FF
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialIridium foil
Name Note0.25mm (0.01 in.) thick, Hard, Temper: as rolled
CAS7439-88-5
Recommended StorageAmbient temperatures
EINECS Number231-095-9