Silver bromide, 99.5%
Silver bromide, 99.5%
Silver bromide, 99.5%
Silver bromide, 99.5%
Thermo Scientific Chemicals
Silver bromide, 99.5%
CAS: 7785-23-1 | AgBr | 187.772 g/mol
Have Questions?
Catalog NumberQuantity
ALF011425.1425 g
Catalog number ALF011425.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver bromide
Melting Point432°C
CAS7785-23-1
ColorYellow
Health Hazard 1H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
View more
Silver bromide is used as the light-sensitive component on ordinary photographic film and plates and also used in I.R. applications and ionic semiconductors.