Ammonium perrhenate(VII), 99+% (metals basis)
Ammonium perrhenate(VII), 99+% (metals basis)
Ammonium perrhenate(VII), 99+% (metals basis)
Ammonium perrhenate(VII), 99+% (metals basis)
Thermo Scientific Chemicals
Ammonium perrhenate(VII), 99+% (metals basis)
CAS: 13598-65-7 | H4NO4Re | 268.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF011410.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium perrhenate(VII)
Melting Point(decomposition)
CAS13598-65-7
ColorWhite
Health Hazard 1Danger
View more
Ammonium perrhenate(VII) is used to synthesis the rehenium(III) organohydrazides which is used as radiopharmaceuticals and also used in the manufacturing of rhenium metal and its additives in superalloys. It is used as catalyst for the synthesis of platinum-rhenium catalysts which used in gasoline.