Silver powder, APS 1-3 micron, 99.9% (metals basis)
Silver powder, APS 1-3 micron, 99.9% (metals basis)
Silver powder, APS 1-3 micron, 99.9% (metals basis)
Silver powder, APS 1-3 micron, 99.9% (metals basis)
Thermo Scientific Chemicals
Silver powder, APS 1-3 micron, 99.9% (metals basis)
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF011405.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver powder
CAS7440-22-4
Recommended StorageAmbient temperatures
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECS Number231-131-3