Iron foil, 0.5mm (0.02in) thick, Puratronic™, 99.992% (metals basis)
Iron foil, 0.5mm (0.02in) thick, Puratronic™, 99.992% (metals basis)
Iron foil, 0.5mm (0.02in) thick, Puratronic™, 99.992% (metals basis)
Iron foil, 0.5mm (0.02in) thick, Puratronic™, 99.992% (metals basis)
Thermo Scientific Chemicals
Iron foil, 0.5mm (0.02in) thick, Puratronic™, 99.992% (metals basis)
CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Catalog NumberQuantity
ALF011379.FI50 x 50 mm
Catalog number ALF011379.FI
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 x 50 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialIron foil
Name Note0.5mm (0.02 in.) thick
CAS7439-89-6
Health Hazard 1H500