Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis)
Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis)
Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis)
Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis)
Thermo Scientific Chemicals
Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis)
Form: Approx. 2.7g/100 x 100mm; Temper: as rolled | CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF011376.GL100 x 400 mm
Catalog number ALF011376.GL
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 x 400 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum
ColorSilver
CAS7429-90-5
AppearanceSilver-colored foil