Aluminum foil, 0.5mm (0.02in) thick, Puratronic™, 99.998% (metals basis)
Aluminum foil, 0.5mm (0.02in) thick, Puratronic™, 99.998% (metals basis)
Aluminum foil, 0.5mm (0.02in) thick, Puratronic™, 99.998% (metals basis)
Aluminum foil, 0.5mm (0.02in) thick, Puratronic™, 99.998% (metals basis)
Thermo Scientific Chemicals
Aluminum foil, 0.5mm (0.02in) thick, Puratronic™, 99.998% (metals basis)
Form: Approx. 3.4g/50 x 50mm; Temper: as rolled | CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
50 x 50 mm
Catalog number ALF011374.GH
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 x 100 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum
ColorSilver
CAS7429-90-5
AppearanceSilver-colored foil