Cadmium powder, -200 mesh, 99.99% (metals basis)
Cadmium powder, -200 mesh, 99.99% (metals basis)
Cadmium powder, -200 mesh, 99.99% (metals basis)
Cadmium powder, -200 mesh, 99.99% (metals basis)
Thermo Scientific Chemicals
Cadmium powder, -200 mesh, 99.99% (metals basis)
CAS: 7440-43-9 | Cd | 112.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF011353.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCadmium powder
ColorWhite
CAS7440-43-9
Health Hazard 1H228-H330-H341-H350-H361fd-H372
Health Hazard 2GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.