Cerium(III) nitrate hexahydrate, REacton™, 99.99% (REO)
Cerium(III) nitrate hexahydrate, REacton™, 99.99% (REO)
Cerium(III) nitrate hexahydrate, REacton™, 99.99% (REO)
Cerium(III) nitrate hexahydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals
Cerium(III) nitrate hexahydrate, REacton™, 99.99% (REO)
CAS: 10294-41-4 | CeH12N3O15 | 434.22 g/mol
Have Questions?
Catalog NumberQuantity
ALF011330.1850 g
Catalog number ALF011330.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium(III) nitrate hexahydrate
Name Notehexahydrate
CAS10294-41-4
Health Hazard 1H272-H315-H318-H350
Health Hazard 3P201-P202-P220-P221-P264b-P271-P280-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P370+P380+P375-P372-P501c
View more
Cerium(III) nitrate hexahydrate is used in the separation of cerium from other rare earths. It plays an important role as a catalyst for hydrolysis of phosphoric acid esters.