Erbium(III) oxide, REacton™, 99.99% (REO)
Erbium(III) oxide, REacton™, 99.99% (REO)
Erbium(III) oxide, REacton™, 99.99% (REO)
Erbium(III) oxide, REacton™, 99.99% (REO)
Thermo Scientific Chemicals
Erbium(III) oxide, REacton™, 99.99% (REO)
CAS: 12061-16-4 | Er2O3 | 382.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALF011309.22
Price (MYR)
3,776.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
3,776.00
EA
Add to cart
Specifications
Chemical Name or MaterialErbium(III) oxide
Melting Point2400°C
CAS12061-16-4
Health Hazard 1H319: Causes serious eye irritation.
Recommended StorageAmbient temperatures
View more
Erbium trioxide is an important rare earth metal used in biomedicine. It is used as a gate dielectric in semi conductor devices since it has a high dielectric constant and a large band gap; as a burnable neutron poison for nuclear fuel and as coloring for glasses. Erbium oxide nanoparticles can be surface modified for distribution into aqueous and non-aqueous media for bioimaging due to its photoluminescence property.