Lanthanum(III) oxide, REacton™, 99.99% (REO)
Lanthanum(III) oxide, REacton™, 99.99% (REO)
Lanthanum(III) oxide, REacton™, 99.99% (REO)
Lanthanum(III) oxide, REacton™, 99.99% (REO)
Thermo Scientific Chemicals
Lanthanum(III) oxide, REacton™, 99.99% (REO)
CAS: 1312-81-8 | La2O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF011264.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Boiling Point4200°C
Chemical Name or MaterialLanthanum(III) oxide
Melting Point2307°C
Name NoteRare Earth Oxides Content
CAS1312-81-8
View more
Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacturing of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics. In addition, it is used as a catalyst to effect certain organic transformations. It is one of the active ingredients and finds application in thermoelectric and piezoelectric materials.