Lutetium(III) oxide, REacton™, 99.99% (REO)
Lutetium(III) oxide, REacton™, 99.99% (REO)
Lutetium(III) oxide, REacton™, 99.99% (REO)
Lutetium(III) oxide, REacton™, 99.99% (REO)
Thermo Scientific Chemicals
Lutetium(III) oxide, REacton™, 99.99% (REO)
CAS: 12032-20-1 | Lu2O3 | 397.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF011256.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLutetium(III) oxide
Melting Point2487°C
Name NoteRare Earth Oxides Content
CAS12032-20-1
ColorWhite
View more
Lutetium(III) oxide serves as a catalyst in a variety of reactions such as polymerization, hydrogenation and alkylation reactions. It is also used in ceramics and glass. In the production of laser crystals, lutetium(III) oxide finds application as an important starting material. Its luminescent properties, a field of interest for researchers and may find potential application in biological fields.