Neodymium(III) oxide, REacton™, 99.9% (REO)
Neodymium(III) oxide, REacton™, 99.9% (REO)
Neodymium(III) oxide, REacton™, 99.9% (REO)
Neodymium(III) oxide, REacton™, 99.9% (REO)
Neodymium(III) oxide, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Neodymium(III) oxide, REacton™, 99.9% (REO)
CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol
Have Questions?
Catalog NumberQuantity
ALF011248.1850 g
Catalog number ALF011248.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNeodymium(III) oxide
Melting Point2272°C
CAS1313-97-9
ColorYellow
Recommended StorageAmbient temperatures
View more
Neodymium(III) oxide is used as a catalyst in polymer chemistry. Glasses doped with neodymium(III) oxide are dichroic in nature and is used in welding goggles and other colored glasses. It finds use in solid-state lasers, carbon arc-light electrodes and enamels. It serves as the starting material for the synthesis of other neodymium(III) compounds.