Praseodymium(III, IV) oxide, 99.5% (REO)
Praseodymium(III, IV) oxide, 99.5% (REO)
Praseodymium(III, IV) oxide, 99.5% (REO)
Praseodymium(III, IV) oxide, 99.5% (REO)
Thermo Scientific Chemicals
Praseodymium(III, IV) oxide, 99.5% (REO)
CAS: 12037-29-5 | O11Pr6 | 1021.44 g/mol
Have Questions?
Catalog NumberQuantity
ALF011234.22100 g
Catalog number ALF011234.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPraseodymium(III, IV) oxide
Melting Point2500°C
CAS12037-29-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
View more
Praseodymium(III, IV) oxide is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.