Samarium(III) chloride hydrate, REacton™, 99.9% (REO)
Samarium(III) chloride hydrate, REacton™, 99.9% (REO)
Samarium(III) chloride hydrate, REacton™, 99.9% (REO)
Samarium(III) chloride hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Samarium(III) chloride hydrate, REacton™, 99.9% (REO)
CAS: 10361-82-7 | Cl3Sm | 256.71 g/mol
Have Questions?
Catalog NumberQuantity
ALF011231.22100 g
Catalog number ALF011231.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSamarium(III) chloride hydrate
Melting Point110°C (in -5H2O)
CAS10361-82-7
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Samarium(III) chloride hydrate is used for the preparation of samarium metal, which finds use as magnets. It is combined with sodium chloride to get a low melting point eutectic mixture. Further, it serves as a precursor for the preparation of other samarium salts and bis(pentamethylcyclopentadienyl)alkylsamarium(III) complexes. In addition, it serves as a catalyst for hydrogenation and hydrosilylation of alkenes.