Samarium(III) oxide, REacton™, 99.9% (REO)
Samarium(III) oxide, REacton™, 99.9% (REO)
Samarium(III) oxide, REacton™, 99.9% (REO)
Samarium(III) oxide, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Samarium(III) oxide, REacton™, 99.9% (REO)
CAS: 12060-58-1 | Sm2O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF011229.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSamarium(III) oxide
Melting Point2300°C
CAS12060-58-1
ColorWhite
Recommended StorageAmbient temperatures
View more
Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. It finds use as a neutron absorber in nuclear power reactors to control the reaction. It is also used as a catalyst for the dehydration of primary alcohols to aldehydes or ketones. It forms samarium perchlorate when treated with perchloric acid, which is used as a reference material for checking the wavelength scale of a spectrophotometer in the range of 200 to 500 nm.