Terbium(III,IV) oxide, REacton™, 99.9% (REO)
Terbium(III,IV) oxide, REacton™, 99.9% (REO)
Terbium(III,IV) oxide, REacton™, 99.9% (REO)
Terbium(III,IV) oxide, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Terbium(III,IV) oxide, REacton™, 99.9% (REO)
CAS: 12037-01-3 | O7Tb4 | 747.69 g/mol
Have Questions?
Catalog NumberQuantity
ALF011207.0910 g
Catalog number ALF011207.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTerbium(III, IV) oxide
Melting Point2340°C
CAS12037-01-3
Recommended StorageAmbient temperatures
Physical FormPowder
View more
Terbium(III,IV) oxide acts as a precursor for the preparation of other terbium compounds. It is used as a redox catalyst in reactions involving oxygen. At high temperatures, it can liberate oxygen. It finds applications in the manufacture of glass, optical and ceramic. Nanoparticles of terbium oxide are used as analytical reagents for the determination of drugs in food.