Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
CAS: 19423-86-0 | Cl3Tm | 275.28 g/mol
Have Questions?
Catalog NumberQuantity
ALF011200.042 g
Catalog number ALF011200.04
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 g
Request bulk or custom format
Specifications
Chemical Name or MaterialThulium(III) chloride hydrate
CAS19423-86-0
ColorGreen
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Thulium(III) chloride hydrate is used in proteomics research. It is also used in the production of glass, phosphors and lasers. Further, it serves as an important dopant for fiber amplifiers. Further, it is also used to prepare thulium(III) oxide.