L-(+)-Ascorbic acid, 98+%
L-(+)-Ascorbic acid, 98+%
L-(+)-Ascorbic acid, 98+%
L-(+)-Ascorbic acid, 98+%
Thermo Scientific Chemicals
L-(+)-Ascorbic acid, 98+%
CAS: 50-81-7 | C6H8O6 | 176.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF011188.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-(+)-Ascorbic acid
CAS50-81-7
Melting Point∽191°C (decomposition)
Recommended StorageAmbient temperatures
Density1.65 g/mL
View more
L-Ascorbic acid is an important nutrient and antioxidant. L-ascorbic acid is frequently used as a processing aid or as a preservative in certain nutrients or food components. It is also utilized in the synthetic polymer industry, in photo processing and in metal technology. Miscellaneous uses have been proposed in cosmetics, fibers, preservation of blood, preservation of cut plants, cleaning agents and in assay reagents.