Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals
Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
CAS: 1310-53-8 | GeO2 | 104.628 g/mol
Have Questions?
Catalog NumberQuantity
ALF011155.1425 g
Catalog number ALF011155.14
Price (MYR)
1,682.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,682.00
EA
Add to cart
Specifications
Chemical Name or MaterialGermanium(IV) oxide
Melting Point1086°C
CAS1310-53-8
ColorWhite
Health Hazard 1H302+H332
View more
Germanium dioxide is used as an optical material for wide-angle lenses and in optical microscope objective lenses. It is used as a catalyst in the preparation of polyethylene terephthalate resin. Further, it is used as a feedstock for the manufacturing of phosphors and semiconductor materials. It is also used in transistors and diodes, and infrared transmitting glass.