Germanium pieces, 2cm (0.8in) and down, Puratronic™, 99.999% (metals basis)
CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Catalog Number
Quantity
ALF011144.06
5 g
Catalog number ALF011144.06
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGermanium pieces
Name NoteElectrical Resistivity: >40 ohm-cm
CAS7440-56-4
Recommended StorageAmbient temperatures
EINECS Number231-164-3