Aluminum wire, 1.0mm (0.04in) dia, annealed, 99+% (metals basis)
Aluminum wire, 1.0mm (0.04in) dia, annealed, 99+% (metals basis)
Aluminum wire, 1.0mm (0.04in) dia, annealed, 99+% (metals basis)
Aluminum wire, 1.0mm (0.04in) dia, annealed, 99+% (metals basis)
Thermo Scientific Chemicals
Aluminum wire, 1.0mm (0.04in) dia, annealed, 99+% (metals basis)
Form: Approx. 2.1g/m; Temper: soft | CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 m
10 m
Catalog number ALF011064.H2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 m
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum
ColorSilver
Name NoteAnnealed
CAS7429-90-5
AppearanceSilver-colored wire