Indium shot, tear drop, 4mm (0.2in), 99.99% (metals basis)
Indium shot, tear drop, 4mm (0.2in), 99.99% (metals basis)
Indium shot, tear drop, 4mm (0.2in), 99.99% (metals basis)
Indium shot, tear drop, 4mm (0.2in), 99.99% (metals basis)
Thermo Scientific Chemicals
Indium shot, tear drop, 4mm (0.2in), 99.99% (metals basis)
CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Catalog NumberQuantity
ALF011027.0910 g
Catalog number ALF011027.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIndium
ColorSilver-White
CAS7440-74-6
AppearanceSilvery-white shot