Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals
Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)
Form: approx. 1.1g/25x200mm | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 200 mm
25 x 1000 mm
Catalog number ALF010950.HV
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 x 200 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper
CAS7440-50-8
AppearanceCopper colored foil
Recommended StorageAmbient temperatures
EINECS Number231-159-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
RUO – Research Use Only