Boron carbide, 90+%
Boron carbide, 90+%
Boron carbide, 90+%
Thermo Scientific Chemicals
Boron carbide, 90+%
CAS: 12069-32-8 | CB4 | 55.25 g/mol
Have Questions?
Catalog NumberQuantity
ALF010922.22100 g
Catalog number ALF010922.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBoron carbide
Melting Point2350°C
CAS12069-32-8
ColorBlack
Health Hazard 1H315-H319-H332-H335
View more
Boron carbide is a hard boron-carbon ceramic material used in tank armor, bulletproof vests, engine sabotage powders, neutron absorber, cutting tools and dies and in brake linings of vehicles. It acts as an antioxidant additives in magnesia-carbon bricks. It is used as a precursor in the production of boron containing materials such as titanium boride.