Holmium(III) oxide, 99.995% (REO), Thermo Scientific™
Holmium(III) oxide, 99.995% (REO), Thermo Scientific™
Holmium(III) oxide, 99.995% (REO), Thermo Scientific™
Holmium(III) oxide, 99.995% (REO), Thermo Scientific™
Thermo Scientific Chemicals
Holmium(III) oxide, 99.995% (REO), Thermo Scientific™
Have Questions?
Catalog number ALF010910.06
Price (MYR)
975.00
Price (MYR)
975.00
EA
Add to cart
Specifications
Chemical Name or MaterialHolmium(III) oxide
Melting Point2360°C
CAS12055-62-8
ColorBeige
Recommended StorageAmbient temperatures
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.