Holmium(III) oxide, REacton™, 99.995% (REO)
Holmium(III) oxide, REacton™, 99.995% (REO)
Holmium(III) oxide, REacton™, 99.995% (REO)
Holmium(III) oxide, REacton™, 99.995% (REO)
Thermo Scientific Chemicals
Holmium(III) oxide, REacton™, 99.995% (REO)
CAS: 12055-62-8 | Ho2O3 | 377.86 g/mol
Have Questions?
Catalog NumberQuantity
ALF010910.031 g
Catalog number ALF010910.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHolmium(III) oxide
Melting Point2360°C
CAS12055-62-8
ColorBeige
Recommended StorageAmbient temperatures
View more
Holmium(III) oxide iacts as colorants which is used for cubic zirconia and glass, as a calibration standard for optical spectrophotometers, as a specialty catalyst, phosphor and a laser material.