Selenous acid, Puratronic™, 99.999% (metals basis)
Selenous acid, Puratronic™, 99.999% (metals basis)
Selenous acid, Puratronic™, 99.999% (metals basis)
Selenous acid, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals
Selenous acid, Puratronic™, 99.999% (metals basis)
CAS: 7783-00-8 | H2O3Se | 128.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF010855.1425 g
Catalog number ALF010855.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSelenous acid
CAS7783-00-8
Melting Point70°C (decomposition)
Health Hazard 1H301+H331-H373
Health Hazard 2GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
View more
Selenous acid is used in steel guns, wherein it changes the color of the steel from silver-grey to blue-grey. It serves as an important component of the Mecke reagent, which is utilized for drug testing. It acts as a source of selenium. It acts as a reagent for alkaloids and an oxidizing agent. Further, it is used in labeling radiopharmaceuticals. It is involved in the organic synthesis for the preparation of glyoxal.