Aluminum rod, 9.5mm (0.37in) dia, Puratronic™, 99.999% (metals basis)
Aluminum rod, 9.5mm (0.37in) dia, Puratronic™, 99.999% (metals basis)
Aluminum rod, 9.5mm (0.37in) dia, Puratronic™, 99.999% (metals basis)
Aluminum rod, 9.5mm (0.37in) dia, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals
Aluminum rod, 9.5mm (0.37in) dia, Puratronic™, 99.999% (metals basis)
CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF010779.BU25 cm
Catalog number ALF010779.BU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 cm
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum rod
Name Note9.5mm (0.37 in.) dia.
CAS7429-90-5
Recommended StorageAmbient temperatures
EINECS Number231-072-3