Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis)
Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis)
Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis)
Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis)
Thermo Scientific Chemicals
Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis)
CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Catalog NumberQuantity
ALF010759.1850 g
Catalog number ALF010759.18
Price (MYR)
624.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
624.00
EA
Add to cart
Specifications
Chemical Name or MaterialZinc
ColorGray
Name Note1-5mm (0.04-0.2 in.)
CAS7440-66-6
AppearanceGray shot