Lithium tetraborate, Puratronic™, 99.998% (metals basis)
Lithium tetraborate, Puratronic™, 99.998% (metals basis)
Lithium tetraborate, Puratronic™, 99.998% (metals basis)
Lithium tetraborate, Puratronic™, 99.998% (metals basis)
Lithium tetraborate, Puratronic™, 99.998% (metals basis)
Thermo Scientific Chemicals
Lithium tetraborate, Puratronic™, 99.998% (metals basis)
CAS: 12007-60-2 | B4Li2O7 | 169.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
Catalog number ALF010744.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium Tetraborate
Melting Point930°C
CAS12007-60-2
ColorWhite
Health Hazard 1H302-H315-H319-H335: Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
View more
Used in the laboratory as LB (Lithium borate) buffer for gel electrophoresis of DNA and RNA. Employed in the borax fusion method to modify mineral powder samples for analysis by WDXRF (Wavelength Dispersive X-Ray Fluorescence) Spectroscopy. Acts as an ingredient in the manufacture of glasses and ceramics. Useful fod dissolving basic oxides such as CaO and MgO. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.