Ammonium chloride, Puratronic™, 99.999% (metals basis)
Ammonium chloride, Puratronic™, 99.999% (metals basis)
Ammonium chloride, Puratronic™, 99.999% (metals basis)
Ammonium chloride, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals
Ammonium chloride, Puratronic™, 99.999% (metals basis)
CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF010632.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium chloride
Melting Point340° (sublimation)
CAS12125-02-9
ColorWhite
Health Hazard 1H302-H319-H335-H373
View more
Ammonium chloride finds its use as a flux in preparing metals for galvanizing, soldering or tin coating. While mainly referred as a nitrogen source fertilizer, it is also used in cleaning tip of soldering iron, hair shampoo as a thickening agent, some batteries as electrolyte, tanning, textile printing, dyeing, and cleaning products. In laboratories it is used to produce cooling baths and for producing buffers. Along with certain metals, it is used for effecting certain reductions in organic synthesis. It can be used as a precursor to ammonium cyanate, which is employed in the Wöhler synthesis of urea. It facilitates several organic transformations in a mild and efficient manner - for example in Ulmann coupling, cross-aldol condensation, Claisen rearrangement, reduction, thia-Michael addition and synthesis of several monocyclic, fused or spiro bicyclic nitrogen heterocycles.