Lithium chloride, anhydrous, 99%
Lithium chloride, anhydrous, 99%
Lithium chloride, anhydrous, 99%
Lithium chloride, anhydrous, 99%
Lithium chloride, anhydrous, 99%
Lithium chloride, anhydrous, 99%
CAS: 7447-41-8 | ClLi | 42.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
Catalog number ALF010515.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Specifications
Boiling Point1325°C to 1360°C
Chemical Name or MaterialLithium chloride, anhydrous
Melting Point605°C
CAS7447-41-8
ColorWhite to Gray
View more
Lithium chloride is useful for the production of lithium metal, and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer, and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere, and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis, and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.