Cobalt powder, 1.6 micron, 99.8% (metals basis)
Cobalt powder, 1.6 micron, 99.8% (metals basis)
Cobalt powder, 1.6 micron, 99.8% (metals basis)
Cobalt powder, 1.6 micron, 99.8% (metals basis)
Cobalt powder, 1.6 micron, 99.8% (metals basis)
Thermo Scientific Chemicals
Cobalt powder, 1.6 micron, 99.8% (metals basis)
CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Catalog number ALF010455.22
Price (MYR)
740.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
740.00
EA
Add to cart
Specifications
Chemical Name or MaterialCobalt powder
ColorBlack
CAS7440-48-4
Health Hazard 1H228-H302-H317-H319-H330-H334-H335-H336-H350i-H360-H373-H500
Health Hazard 2GHS H Statement
H228-H334-H317
Flammable solid.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.