Zinc foil, 0.25mm (0.01in) thick, 30cm (12in) wide, 99.98% (metals basis)
Zinc foil, 0.25mm (0.01in) thick, 30cm (12in) wide, 99.98% (metals basis)
Zinc foil, 0.25mm (0.01in) thick, 30cm (12in) wide, 99.98% (metals basis)
Zinc foil, 0.25mm (0.01in) thick, 30cm (12in) wide, 99.98% (metals basis)
Zinc foil, 0.25mm (0.01in) thick, 30cm (12in) wide, 99.98% (metals basis)
Thermo Scientific Chemicals
Zinc foil, 0.25mm (0.01in) thick, 30cm (12in) wide, 99.98% (metals basis)
Form: approx. 53g/30x10cm | CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Catalog NumberQuantity
ALF010436.BW50 cm
Catalog number ALF010436.BW
Price (MYR)
529.00
EA
Add to cart
Quantity:
50 cm
Request bulk or custom format
Price (MYR)
529.00
EA
Add to cart
Specifications
Chemical Name or MaterialZinc
ColorGray
CAS7440-66-6
AppearanceGray foil
Density7.14g/cm3