Tungsten wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)
Tungsten wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)
Tungsten wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)
Tungsten wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)
Thermo Scientific Chemicals
Tungsten wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)
CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Catalog NumberQuantity
ALF010409.G55 m
Catalog number ALF010409.G5
Price (MYR)
597.00
EA
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (MYR)
597.00
EA
Add to cart
Specifications
Chemical Name or MaterialTungsten
CAS7440-33-7
AppearanceSilver Gray wire
Density19.3g/cm3