Tungsten powder, APS 1-5 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Tungsten powder, APS 1-5 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Tungsten powder, APS 1-5 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Tungsten powder, APS 1-5 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tungsten powder, APS 1-5 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF010400.36500 g
Catalog number ALF010400.36
Price (MYR)
1,353.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,353.00
EA
Add to cart
Specifications
Chemical Name or MaterialTungsten powder
ColorGray
CAS7440-33-7
Health Hazard 1H228-H500
Health Hazard 2GHS H Statement
H228
Flammable solid.